Flat flange lids, centre neck
bride de laboratoire DN 200, 1 col, NS 45/40 (centre), 165 mm
N° de catalogue LENZ06100506
Fournisseur: Lenz Laborglas
Aller à :
Product Details & Documents
DURAN®, Borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Flange lids for reaction vessels, with laboratory flange LF.
Spécifications
- Type:1 col
- Avec:bride de laboratoire DN 200
- Col:NS 45/40 (centre)
- Hauteur totale:165 mm