Flat flange lids, centre neck
bride de laboratoire DN 100, 1 col, NS 45/40 (centre), 110 mm
Catalog # LENZ06100206
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
DURAN®, Borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Flange lids for reaction vessels, with laboratory flange LF.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specifications
- Type:1 col
- Avec:bride de laboratoire DN 100
- Col:NS 45/40 (centre)
- Hauteur totale:110 mm