Extraction thimbles
Extraction thimbles
250 ml
N° de catalogue LENZ05405249
Fournisseur:   Lenz Laborglas
Extraction thimbles
N° de catalogue LENZ05405249
Fournisseur:  Lenz Laborglas
Extraction thimbles
Extraction thimbles
250 ml
Prix
62,10 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
Glass tubes with sintered filter disc (porosity: 0), for extractors acc. to Thielepape.
Documents
Spécifications
  • Capacité:
    250 ml
  • Seq:
    03