Extraction thimbles
Extraction thimbles
30 ml
Catalog # LENZ05405225
Supplier:   Lenz Laborglas
Extraction thimbles
Catalog # LENZ05405225
Supplier:  Lenz Laborglas
Extraction thimbles
Extraction thimbles
30 ml
Price
47,70 €
Log in for availability
1 * 1 ST
Product Details & Documents
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
Glass tubes with sintered filter disc (porosity: 0), for extractors acc. to Thielepape.
Documents
Specifications
  • Capacité:
    30 ml
  • Seq:
    01