Septum caps, Nalgene®
24 - 415
N° de catalogue NALG342178-0240
Fournisseur: Thermo Fisher Scientific
Septum caps, Nalgene®
N° de catalogue NALG342178-0240
Fournisseur: Thermo Fisher Scientific
Aller à :
Product Details & Documents
HDPE with silicone/PTFE septum.
- Sterile 10⁻⁶ SAL
- No adhesives used to attach septum to high density polyethylene closure
- Non-cytotoxic and non-pyrogenic
- Dual septum material for improved gas barrier
Spécifications
- Taille du bouchon:24 - 415
- Cat. No.:NALG342178-0240
- Certification environnementale:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact