Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Flexible distributors for bottles, BOLA

Flexible distributors for bottles, BOLA

deux raccords de tuyaux, 7 - 11 mm
N° de catalogue 201-1087
Fournisseur:   Bohlender
Flexible distributors for bottles, BOLA
N° de catalogue 201-1087
Fournisseur:  Bohlender
Flexible distributors for bottles, BOLA
Flexible distributors for bottles, BOLA

deux raccords de tuyaux, 7 - 11 mm

Prix
84,60 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents

PP screw cap, green, distributor body and tubing connectors made from PP

  • Very good chemical resistance
  • Temperature resistance from 0 to +110 °C
  • Suitable for vacuum

Drawing or inserting liquids. For elastic tubing (e.g. Viton®, Tygon®, silicone). Bent hose connectors on upper side, straight hose connectors on lower side.

Spécifications
  • Tube Ø int.:
    7 - 11 mm
  • Désignation:
    deux raccords de tuyaux