Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
 

Microtubes à centrifuger

2,0 ml, naturel, With attached sealing caps, Non graduated, conical bottom, non sterile
N° de catalogue RATI5615101
Fournisseur:   RATIOLAB
Microtubes à centrifuger
N° de catalogue RATI5615101
Fournisseur:  RATIOLAB
Microtubes à centrifuger
Microtubes à centrifuger

2,0 ml, naturel, With attached sealing caps, Non graduated, conical bottom, non sterile

Prix
20,70 €
Connexion Pour la disponibilité
1 * 1.000 ST
Product Details & Documents

PP, translucent.

  • Autoclavable with opened cap at 121 °C, 20 min
  • Natural colour or different colours for comfortable identification
  • Tight sealing cap
  • Safe and easy to be opened and closed

With or without flat cap.

Spécifications
  • Type:
    With attached sealing caps
  • Version:
    Non graduated, conical bottom, non sterile
  • Capacité:
    2,0 ml
  • Couleur:
    naturel
  • Seq:
    08