D-(+)-Cellobiose ≥98%
Glass bottle for solids, 100 g
Fournisseur: Thermo Fisher Scientific
D-(+)-Cellobiose ≥98%
N° de catalogue A14553.22
Fournisseur: Thermo Fisher Scientific
Numéro CAS: 528-50-7
Aller à :
Product Details & Documents
Purity: 98+% CAS No.: 528-50-7 Molecular Formula: C12H22O11 Formula Weight: 342.30 Physical Form: Powder Appearance: White Melting Point: 234° dec. Beilstein Reference: 93795 Merck Reference: 14,1961 Optical Rotation: +34° (c=10 in water, 15h) MDL No.: MFCD00136034
Synonymes: β-D-Glc-(1→4)-D-Glc, 4-β-D-glucopyranosyl-D-glueopyranose, Cellose, β-Cellobiose, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal, 4-O-β-D-Glucopyranosyl-D-glucopyranose
- Numéro CAS:528-50-7
- Formule:C₁₂H₂₂O₁₁
- Numéro MDL:MFCD00136034
- Poids moléculaire:342.30 g/mol
- Index Merck:14,1961
- Point de fusion:234 °C (déc.)
- Température de stockage:Température ambiante
- EINECS:208-436-5
Documents
Spécifications
Spécifications
- Type de conditionnement:Glass bottle for solids
- Cdt:100 g
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



