Acide 3,4-dihydroxybenzoïque ≥97%
Flacon verre pour solides, 500 g
Supplier: Thermo Fisher Scientific
Acide 3,4-dihydroxybenzoïque ≥97%
Catalog # B24016.36
Supplier: Thermo Fisher Scientific
CAS Number: 99-50-3
Jump to:
Product Details & Documents
Purity: 97%
CAS No.: 99-50-3
Molecular Formula: C7H6O4
Formula Weight: 154.12
Melting Point: ca 202° dec.
Beilstein Reference: 1448841
Merck Reference: 14,7894
MDL No.: MFCD00002509
Synonyms: Protocatechuic acid
- CAS Number:99-50-3
- Formula:C₇H₆O₄
- MDL Number:MFCD00002509
- Molecular Weight:154.12 g/mol
- Merck Index:14,7894
- Boiling Pt:125…135 °C (0,001 mmHg)
- Melting Pt:∼202 °C (déc.)
- Density:1,5415 g/cm³ (20 °C)
- Storage Temperature:Température ambiante
- EINECS:202-760-0
Documents
Specifications
Specifications
- Type de conditionnement:Flacon verre pour solides
- Cdt:500 g



