Bathocuproin ≥98%
Flacon verre pour solides, 5 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 98% CAS No.: 4733-39-5 Molecular Formula: C26H20N2 Formula Weight: 360.46 Melting Point: 279-285° Beilstein Reference: 306714
Synonyms: 2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline
- CAS Number:4733-39-5
- Formula:C₂₆H₂₀N₂
- MDL Number:MFCD00004972
- Molecular Weight:360.46 g/mol
- Melting Pt:279…285 °C
- Storage Temperature:Température ambiante
- EINECS:225-240-5
Documents
Specifications
Specifications
- Type de conditionnement:Flacon verre pour solides
- Cdt:5 g



