n-Hexadécane ≥99%
Product Details & Documents
Hexadecane is use in the recovery of valuable amines by means of distillation from aqueous alkanolamine solutions, which are extensively used in the gas-processing industries.
Purity: 99%
CAS No.: 544-76-3
Molecular Formula: CH3(CH2)14CH3
Formula Weight: 226.45
Flash Point: 135°(275°F)
Physical Form: Liquid
Density: 0.773
Refractive Index: 1.4345
Appearance: Colourless
Melting Point: 18°
Boiling Point: 287°
Solubility: This product is soluble in acetone, alcohol, ether. It is insoluble in water.
Beilstein Reference: 1736592
MDL No.: MFCD00008998
Hexadecane is use in the recovery of valuable amines by means of distillation from aqueous alkanolamine solutions, which are extensively used in the gas-processing industries. *Majid Abedinzadegan Abdi and Axel Meisen. Mutual Solubility of Hexadecane with Diethanolamine and with Bis(hydroxyethyl)piperazine J. Chem. Eng. Data, 1998, 43 (2), pp 138-142
- Numéro CAS:544-76-3
- Formule:H₃C(CH₂)₁₄CH₃
- Numéro MDL:MFCD00008998
- Poids moléculaire:226.45 g/mol
- Point d'ébullition:287 °C
- Point de fusion:18 °C
- Densité:0,773
- Point d'éclair:135 °C (275 °F)
- Température de stockage:Température ambiante
- EINECS:208-878-9
Documents
Certificats
Rechercher par numéro de lotOrganic Building Blocks and Intermediates
Notes d'application
Spécifications
Spécifications
- Type de conditionnement:Flacon Verre
- Cdt:500 mL
- Cat. No.:A10322.AP



