Dichlorométhane ≥99% stabilisé
Flacon Verre, 2.500 mL
Supplier: Thermo Fisher Scientific
Dichlorométhane ≥99% stabilisé
Catalog # L13089.0F
Supplier: Thermo Fisher Scientific
CAS Number: 75-09-2
Jump to:
Product Details & Documents
Stabilized with ca. 50 ppm amylene
Purity: 99+%
CAS No.: 75-09-2
Molecular Formula: CH2Cl2
Formula Weight: 84.93
Refractive Index: 1.4244
Odour: Sweet
Boiling Point: 39-40°
Beilstein Reference: 1730800
Merck Reference: 14,6063
UN No.: 1593
MDL No.: MFCD00000881
Synonyms: Methylene chloride, DCM
- CAS Number:75-09-2
- Formula:CH₂Cl₂
- MDL Number:MFCD00000881
- Molecular Weight:84.93 g/mol
- Merck Index:14,6063
- Boiling Pt:39…40 °C
- Melting Pt:–95 °C
- Density:1,325
- Storage Temperature:Température ambiante
- EINECS:200-838-9
- ADR:6.1,III
- :stabilisé avec 2-méthyl-2-butène (iso-amylène) 50 ppm
- UN:1593
Documents
Specifications
Specifications
- Type de conditionnement:Flacon Verre
- Cdt:2.500 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



