Mailing boxes
Mailing boxes with enclosed foam bag for 1 - 10 containers, 220×170×40 mm
N° de catalogue SARS95.902
Marketsource
Fournisseur: SARSTEDT
Aller à :
Product Details & Documents
Mailing boxes, made of cardboard, for safe transport of diagnostic specimen in containers.
- With printed label
- Lightweight
- Sturdy
Post-transport packaging for diagnostic samples in accordance with packaging regulation P 650, oversize letter, structural authorisation, flat design.
Spécifications
- Désignation:Mailing boxes with enclosed foam bag for 1 - 10 containers
- Seq:03
- l×p×h:220×170×40 mm