Screw caps for micro tubes
Screw caps for micro tubes, 10 colours, assortiment
Catalog # SARS65.716.999
Marketsource
Supplier: SARSTEDT
Jump to:
Product Details & Documents
PP, coloured.
- Excellent chemical resistance
- Excellent temperature resistance
Screw cap with O-ring seal.
Specifications
- Couleur:assortiment
- Désignation:Screw caps for micro tubes, 10 colours
- Seq:01