Mailing boxes
Mailing boxes with integral restriants for 1 - 4 containers, 198×107×38 mm
N° de catalogue SARS95.900
Marketsource
Fournisseur: SARSTEDT
Aller à :
Product Details & Documents
Mailing boxes, made of cardboard, for safe transport of diagnostic specimen in containers.
- With printed label
- Lightweight
- Sturdy
Post-transport packaging for diagnostic samples in accordance with packaging regulation P 650, oversize letter, structural authorisation, flat design.
Spécifications
- Désignation:Mailing boxes with integral restriants for 1 - 4 containers
- Seq:01
- l×p×h:198×107×38 mm