Pressure Bulbs
Atomiser bulb with suction valve, red-brown, size 3, PVC, 65
N° de catalogue DENE4250003
Fournisseur: DEUTSCH NEUMANN
Aller à :
Product Details & Documents
Natural rubber, chloroprene or PVC.
Used for filling automatic burettes, Pellet pattern (ball set with net) or atomisers (bulb).
Spécifications
- Capacité:65
- Désignation:Atomiser bulb with suction valve, red-brown, size 3, PVC
- Seq:2