Microplates, opaque OptiPlates™
OptiiPlate™-96 F microplates, 96, noir, Case of 50
N° de catalogue PERK6055270
Marketsource
Fournisseur: PerkinElmer
Aller à :
Product Details & Documents
These plates are used for general isotopic, fluorescence, LANCE™, [FP]², AlphaScreen™ or luminescence assays.
- Made of polystyrene
Spécifications
- Nb de puits:96
- Emballage:Case of 50
- Couleur:noir
- Désignation:OptiiPlate™-96 F microplates
- Seq:01