Screw caps
GL 32
N° de catalogue VITL83312
Fournisseur: VITLAB
Aller à :
Product Details & Documents
Made of PP, translucent.
Caps are autoclavable at 121 °C (2 bar) according to DIN EN 285.
Spécifications
- Filetage:GL 32
Made of PP, translucent.
Caps are autoclavable at 121 °C (2 bar) according to DIN EN 285.