Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
 

Overalls, Proshield® 8 Proper, model CCF5

gris, XL
  DUPOCCF5SGY00/XL
 :   DuPont
Overalls, Proshield® 8 Proper, model CCF5
  DUPOCCF5SGY00/XL
 :  DuPont
Overalls, Proshield® 8 Proper, model CCF5
Overalls, Proshield® 8 Proper, model CCF5

gris, XL

 
954,00 €
   
1 * 100 ST
 

ProShield® Proper overalls are made of a proprietary DuPont non woven PE fabric (55 g/m²) that makes them incredibly tough, lightweight and resistant to tears and punctures.

  • Mandarin collar, nylon zipper with flap
  • Two thigh pockets
  • Stitched seams
  • Wrist and back elastication for good fit, open ankles (not elasticated)
  • Antistatically treated for comfort (not acc. EN 1149-5)
  • Can be re-used several times and may also be washed up to seven times

The overalls are comfortable because the material allows the passage of air and water vapour. They are ideal workwear for do-it-yourself, general maintenance and cleaning, manufacturing and other non hazardous applications as well as a semi re-usable garment for visitors.

Colour: Grey

 : Cat. I

 
  • Taille de confection:
    XL
  • Couleur:
    gris
  • Seq:
    04
  • Cat. No.:
    DUPOCCF5SGY00/XL
  • Anti-statique:
    +
  • Poids:
    55 g/m²
  • Applications des tenues:
    À usage universel|Alimentaire
  • Modèle:
    CCF5