Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Aluminium sulfate 30% en solution aqueuse

Aluminium sulfate 30% en solution aqueuse

Flacon Plastique, 20 L
Catalog # KRAF01636.7000
Corrosive
Supplier:   Bernd Kraft
Aluminium sulfate 30% en solution aqueuse
Catalog # KRAF01636.7000
Supplier:  Bernd Kraft
CAS Number:  10043-01-3
Aluminium sulfate 30% en solution aqueuse

Flacon Plastique, 20 L

CAS Number:  10043-01-3
Price
392,00 €
Log in for availability
1 * 20 L
Product Details & Documents
Synonyms:  Aluminum sulphate
  • CAS Number:10043-01-3
  • Formula:Al₂O₁₂S₃
  • MDL Number:MFCD00003423
  • Molecular Weight:342.15 g/mol
  • Boiling Pt:∼100 °C (1013 hPa)
  • Density:∼1,2 g/cm³ (20 °C)
  • EINECS:233-135-0
Specifications

Specifications

  • Type de conditionnement:
    Flacon Plastique
  • Cdt:
    20 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.