Solid phase extraction cartridges, Bond Elut™ Florisil
Bond Elut™ Jr, Florisil, 200 µm, 500 mg
N° de catalogue VARI12162050B
Fournisseur: VARIAN
Solid phase extraction cartridges, Bond Elut™ Florisil
N° de catalogue VARI12162050B
Fournisseur: VARIAN
Aller à :
Product Details & Documents
Florisil is a magnesia-loaded silica gel, extremely polar in nature and ideal for the isolation of polar compounds from non-polar matrices.
- Pesticide residue (PR) grade
- Fast flow, ideal for viscous samples
The larger particle size of the sorbent enables flow for large sample volumes and is therefore an attractive alternative to silica if the sample matrix is particularly viscous.
Spécifications
- Granulométrie:200 µm
- Désignation:Bond Elut™ Jr
- Seq:14
- Poids du lit:500 mg
- Greffage:Florisil