Particle traps for use with PLOT columns
Particle traps for use with PLOT columns
Particle trap connectors for PoraPLOT columns, fused silica, 0,53 mm ID
N° de catalogue VARICP4789
Fournisseur:   VARIAN
Particle traps for use with PLOT columns
N° de catalogue VARICP4789
Fournisseur:  VARIAN
Particle traps for use with PLOT columns
Particle trap connectors for PoraPLOT columns, fused silica, 0,53 mm ID
Prix
227,00 €
Connexion Pour la disponibilité
1 * 10 ST
Product Details & Documents
Though highly stabilised, it is impossible to guarantee that no particles will dislodge from the column wall. When used in valve-switching applications, the use of a particle trap can prevent scarring of the rotors.
Spécifications
  • Désignation:
    Particle trap connectors for PoraPLOT columns, fused silica, 0,53 mm ID
  • Seq:
    07