Solid phase extraction cartridges, Bond Elut™ SI
Solid phase extraction cartridges, Bond Elut™ SI
120
N° de catalogue VARI14113036
Fournisseur:   VARIAN
Solid phase extraction cartridges, Bond Elut™ SI
N° de catalogue VARI14113036
Fournisseur:  VARIAN
Solid phase extraction cartridges, Bond Elut™ SI
120
Prix
252,00 €
Connexion Pour la disponibilité
1 * 50 ST
Product Details & Documents
Native silica is generally regarded as the most polar SPE sorbent available. Bond Elut™ SI is particularly effective at separating compounds with a very similar structure. Applying the analytes in a non-polar solvent, then increasing the solvent polarity by increasing the concentration of a polar modifier, such as THF or ethyl acetate, delivers effective separations.
  • Highly polar phase retains polar molecules from non-polar matrices
  • High purity silica
  • Separate compounds with very similar structures
Spécifications
  • Granulométrie:
    120