Inlet septa
Inlet septa
Septa, general purpose, red, 11 mm
N° de catalogue HEWL5181-3383-100
Fournisseur:   Agilent
Inlet septa
N° de catalogue HEWL5181-3383-100
Fournisseur:  Agilent
Inlet septa
Inlet septa
Septa, general purpose, red, 11 mm
Prix
156,00 €
Connexion Pour la disponibilité
1 * 100 ST
Product Details & Documents
These septa are available for a variety of applications and have different upper temperature limits. The lower temperature septa are usually softer, seal better and can withstand more punctures (injections) than their high temperature counterparts.
  • Made up low bleed silicone rubber formulations
  • Efficient by using sharp syringe needles
  • Septa should be replaced to avoid leaks and sample loss
  • To be used with recommended temperature range
Spécifications
  • Ø:
    11 mm
  • Désignation:
    Septa, general purpose, red