Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Gadoteridol, Reference Standards

Gadoteridol, Reference Standards

Gadoteridol Related Compound C, USP reference standard, Vial, 50 mg
N° de catalogue 1287664.
Fournisseur:   USP
Gadoteridol, Reference Standards
N° de catalogue 1287664.
Fournisseur:  USP
Numéro CAS:  
Gadoteridol, Reference Standards

Gadoteridol Related Compound C, USP reference standard, Vial, 50 mg

Numéro CAS:  
Prix
1 140,00 €
Connexion Pour la disponibilité
1 * 50 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Spécifications

Spécifications

  • Type de conditionnement:
    Vial
  • Cdt:
    50 mg
  • Normes:
    USP reference standard
  • Désignation:
    Gadoteridol Related Compound C
  • Cat. No.:
    1287664.
  • N ° CAS.:
    220182-19-4
  • Désignation:
    Gadoteridol Related Compound C
  • Synonyme(s):
    1,4,7,10-Tetraaza-11-oxo-bicyclo[8.2.2]tetradecane-4,7-diacetic acid

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.