Racks for freezer
20 (4×5), 50 mm, 566×140×274 mm
N° de catalogue RATI5400003
Fournisseur: RATIOLAB
Aller à :
Product Details & Documents
For the storing of cryoboxes in freezer cabinets and chests.
- Robust construction
- Handles for convenient transport
- Made of stainless steel with surfaces for perfect cleaning
Delivery information: Cryo racks are supplied with shelf sizes for any standard cryoboxes.
Spécifications
- Compartiments:20 (4×5)
- l×p×h ext.:566×140×274 mm
- Seq:06
- Hauteur de cryoboîte:50 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



