Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Internal reducing union, zero dead volume, Valco®

Internal reducing union, zero dead volume, Valco®

Internal reducing union, Acier inoxydable, 0,25 mm
N° de catalogue REST20151
Fournisseur:   Restek
Internal reducing union, zero dead volume, Valco®
N° de catalogue REST20151
Fournisseur:  Restek
Internal reducing union, zero dead volume, Valco®
Internal reducing union, zero dead volume, Valco®

Internal reducing union, Acier inoxydable, 0,25 mm

Prix
95,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents

Internal reducing union used to connect two sizes of tubing, using zero-dead-volume fittings on each end. Suitable for 1,6 to 3,2 mm OD tubing.

Delivery information: Supplied with stainless steel ferrules.

Spécifications
  • Ø trou:
    0,25 mm
  • Désignation:
    Internal reducing union
  • Matière:
    Acier inoxydable
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.