Flow controllers, Veriflo
0,5 – 2,0 ml/min, Acier inoxydable traité Siltek®, Flow controller, 0,02 mm
N° de catalogue REST24232
Fournisseur: Restek
Aller à :
Product Details & Documents
Veriflo controllers can be used with passive air samplers in maintaining a constant flow rate.
Entire sample flow path is made of stainless steel or siltek® treated for maximum inertness.
Spécifications
- Ø:0,02 mm
- Débit maximal:0,5 – 2,0 ml/min
- Désignation:Flow controller
- Matière:Acier inoxydable traité Siltek®
- Seq:04