Passive air sampling kits
Replacement frit filters, 2 µm, —, Acier inoxydable
N° de catalogue REST24170
Fournisseur: Restek
Aller à :
Product Details & Documents
This unique grab sampler minimises connections and leaks and houses a critical orifice in the tee fitting.
- Different orifice sizes for sampling from four minutes to ten hours
- Compression sample inlet allows easy connection
- Canister valve connected to a sample port tubing
- Stainless steel tee with orifice and vaccum gauge
- Frit filter for insertion into a compression sample inlet
Specifically designed for soil gas sampling by allowing the connection of tubing coming from the soil gas sample port.
Ordering information: Canisters and replacement tees and orifices can be ordered separately.
Spécifications
- Désignation:Replacement frit filters, 2 µm
- Matière:Acier inoxydable
- Seq:901
- Flux d´air:—