Acétonitrile
Flacon Verre, 2,5 L
Catalog # ROTH8825.2
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
- CAS Number:75-05-8
- Formula:H₃CCN
- MDL Number:MFCD00001878
- Molecular Weight:41.05 g/mol
- Merck Index:13,00071
- Boiling Pt:81,6 °C (1013 hPa)
- Melting Pt:–45,7 °C
- Density:0,782 g/cm³ (20 °C)
- Flash Pt:2 °C (coupe fermée)
- Storage Temperature:Température ambiante
- EINECS:200-835-2
- UN:1648
- ADR:3,II
- REACH:01-2119471307-38
Specifications
Specifications
- Type de conditionnement:Flacon Verre
- Cdt:2,5 L



