Dichlorométhane
10 L
Catalog # ROTH8424.3
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Methylene chloride, DCM
- CAS Number:75-09-2
- Formula:CH₂Cl₂
- MDL Number:MFCD00000881
- Molecular Weight:84.93 g/mol
- Merck Index:12,06140
- Boiling Pt:39,8 °C (1013 hPa)
- Melting Pt:–95 °C
- Density:1,322 g/cm³ (20 °C)
- Storage Temperature:Température ambiante
- EINECS:200-838-9
- UN:1593
- ADR:6.1,III
Specifications
Specifications
- Cdt:10 L