Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
 

1-Méthylimidazole

Glass bottle, 1 L
Catalog # ROTHN353.2
Corrosive
Irritant
Marketsource
Supplier:   Roth Carl
1-Méthylimidazole
Catalog # ROTHN353.2
Supplier:  Roth Carl
Marketsource
CAS Number:  616-47-7
1-Méthylimidazole

Glass bottle, 1 L

CAS Number:  616-47-7
Price
142,50 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  N-Methylimidazole, 1-Methyl-1H-imidazole, N-Methyl-1H-imidazole, NMI
  • CAS Number:616-47-7
  • Formula:C₄H₆N₂
  • MDL Number:MFCD00005292
  • Molecular Weight:82.11 g/mol
  • Boiling Pt:198 °C (1013 hPa)
  • Melting Pt:–2 °C
  • Density:1,03 g/cm³ (20 °C)
  • Flash Pt:92 °C
  • Storage Temperature:Température ambiante
  • EINECS:210-484-7
  • UN:3267
  • ADR:8,II
Specifications

Specifications

  • Type de conditionnement:
    Glass bottle
  • Cdt:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.