D-(+)-Cellobiose
Glass bottle for solids, 5 g
N° de catalogue ROTH5840.1
Marketsource
Fournisseur: Roth Carl
Aller à :
Product Details & Documents
Synonymes: β-D-Glc-(1→4)-D-Glc, 4-β-D-glucopyranosyl-D-glueopyranose, Cellose, β-Cellobiose, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal, 4-O-β-D-Glucopyranosyl-D-glucopyranose
- Numéro CAS:528-50-7
- Formule:C₁₂H₂₂O₁₁
- Numéro MDL:MFCD00136034
- Poids moléculaire:342.30 g/mol
- Température de stockage:Température ambiante
- EINECS:208-436-5
Spécifications
Spécifications
- Type de conditionnement:Glass bottle for solids
- Cdt:5 g
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



