Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Acétonitrile

Acétonitrile

Flacon Verre, 2,5 L
Catalog # ROTH4722.2
Flammable
Irritant
Marketsource
Supplier:   Roth Carl
Acétonitrile
Catalog # ROTH4722.2
Supplier:  Roth Carl
Marketsource
CAS Number:  75-05-8
Acétonitrile
Acétonitrile

Flacon Verre, 2,5 L

CAS Number:  75-05-8
Price
115,00 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
  • CAS Number:75-05-8
  • Formula:H₃CCN
  • MDL Number:MFCD00001878
  • Molecular Weight:41.05 g/mol
  • Merck Index:13,00071
  • Boiling Pt:81,6 °C (1013 hPa)
  • Melting Pt:–45,7 °C
  • Density:0,782 g/cm³ (20 °C)
  • Flash Pt:2 °C (coupe fermée)
  • Storage Temperature:Température ambiante
  • EINECS:200-835-2
  • UN:1648
  • ADR:3,II
  • REACH:01-2119471307-38
Specifications

Specifications

  • Type de conditionnement:
    Flacon Verre
  • Cdt:
    2,5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.