ATC probes, Eutech
ATC probes, Eutech
ATC probe, 1 m cable, CyberScan pH 300 / 310
Catalog # EUTEECPHWPTEM01W
Supplier:   EUTECH
ATC probes, Eutech
Catalog # EUTEECPHWPTEM01W
Supplier:  EUTECH
ATC probes, Eutech
ATC probes, Eutech
ATC probe, 1 m cable, CyberScan pH 300 / 310
Price
113,30 €
Log in for availability
1 * 1 ST
Product Details & Documents
ATC electrodes for use with Eutech pH 5+, pH 6+ and ION 6+ meters or for use with CyberScan pH 300 and 310 meters.
Specifications
  • Pour:
    CyberScan pH 300 / 310
  • Désignation:
    ATC probe, 1 m cable