Biochemical ID test systems
Biochemical ID test systems
Gram Negative Biochemical ID Test System for the identification of Enterobacteriaceae and other non fastidious, oxidase negative, gram negative bacilli, 30 Tests
N° de catalogue 709-0266
Fournisseur:   TECRA
Biochemical ID test systems
N° de catalogue 709-0266
Fournisseur:  TECRA
Numéro CAS:  
Biochemical ID test systems
Biochemical ID test systems
Gram Negative Biochemical ID Test System for the identification of Enterobacteriaceae and other non fastidious, oxidase negative, gram negative bacilli, 30 Tests
Numéro CAS:  
Prix
Demander un prix
Connexion Pour la disponibilité
1 * 30 Tests
Product Details & Documents
ID kits can be used to identify pure cultures but are also useful for the identification and confirmation of isolates that have produced presumptive positive results in any of TECRA’s Salmonella or Listeria rapid screening kits. The ID kits employ just 12 standardised microwell substrates. They are simple to use, with no assembly required, single colony inoculation and non-toxic reagents. The Identification System Software can be used to provide a reliable identification.
  • The kits are conveniently sized and contain all the reagents required
  • ID kits can save time and money, eliminating the need to make up numerous types of biochemical test media
  • Greater accuracy and reliability
Spécifications
Spécifications
  • Cdt:
    30 Tests
  • Désignation:
    Gram Negative Biochemical ID Test System for the identification of Enterobacteriaceae and other non fastidious, oxidase negative, gram negative bacilli