Replacement inlet seals
Replacement inlet seals
Inlet seal, used with single column, Siltek®-treated, 0,8 mm Ø int.
N° de catalogue REST21320
Fournisseur:   Restek
Replacement inlet seals
N° de catalogue REST21320
Fournisseur:  Restek
Replacement inlet seals
Replacement inlet seals
Inlet seal, used with single column, Siltek®-treated, 0,8 mm Ø int.
Prix
456,00 €
Connexion Pour la disponibilité
1 * 10 ST
Product Details & Documents
Inlet seals are used at the base of the Agilent 5890/6890/7890 GC injection port, which contacts the sample and should be changed frequently to prevent adsorption of active compounds. In addition, septum fragments and sample residue accumulate on the disk surface, requiring disk replacement.
Spécifications
  • Désignation:
    Inlet seal, used with single column, Siltek®-treated, 0,8 mm Ø int.
Accessories
Informations sur le produit
  • N° de catalogue
    Désignation
    Disponibilité
    Prix
  • REST21710
    Replacement inlet seal washers
    Connexion Pour la disponibilité
    1 * 15 ST - 17,60 €