Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Culture flasks, Erlenmeyer shape, BIOGEN

Culture flasks, Erlenmeyer shape, BIOGEN

500 ml, 105 mm, 105 mm, 38 mm
  WITG5.507.106
Culture flasks, Erlenmeyer shape, BIOGEN
  WITG5.507.106
Culture flasks, Erlenmeyer shape, BIOGEN
Culture flasks, Erlenmeyer shape, BIOGEN

500 ml, 105 mm, 105 mm, 38 mm

 
44,00 €
   
1 * 1 ST
 

Culture flasks, made of borosilicate glass, Erlenmeyer shape, designed for orbital shakers.

  • Straight neck for metal caps
  • Triple baffled shake flask
  • Excellent chemical and temperature resistance
 
  • Ø ext.:
    105 mm
  • Capacité:
    500 ml
  • Hauteur:
    105 mm
  • Ø ext. col:
    38 mm