Plongeurs de tubes
Tubing sinkers, fits tubing with inner Ø 8-10 mm
Supplier: Bel-Art Products, a Part of SP
Jump to:
Product Details & Documents
These Teflon® sinkers help to keep tubing at the bottom of the container.
- Ideal for use with pumps and dispensers
- Hose barb ends for simple tubing connection
- Solvent resistant and autoclavable
Documents
Certificates
Search by Lot NumberSpecifications
- Désignation:Tubing sinkers, fits tubing with inner Ø 8-10 mm
- Seq:03