Hardness tablets
Hardness tablets
T-Hardness speed test tablets, AF445 Minikit, 100
Catalog # LOVI515590BT
Supplier:   Lovibond
Hardness tablets
Catalog # LOVI515590BT
Supplier:  Lovibond
CAS Number:  
Hardness tablets
T-Hardness speed test tablets, AF445 Minikit, 100
CAS Number:  
Price
41,40 €
Log in for availability
1 * 100 ST
Product Details & Documents
The speed test is based on reverse titration. After adding a reagent tablet to a calibrated test tube, the water sample is added slowly until the colour of the solution changes (e.g. from red to blue). The user can then obtain the result from the liquid level.
A Yes/No test tells the user whether a specific ingredient is present in the water and/or if its concentration is higher or lower than a defined level.
A Yes/No test tells the user whether a specific ingredient is present in the water and/or if its concentration is higher or lower than a defined level.
Documents
Specifications
Specifications
  • Cdt:
    100
  • Désignation:
    T-Hardness speed test tablets
  • Pour appareil:
    AF445 Minikit
Specifications