Drying tubes, U-shaped
Drying tubes, U-shaped
12/21 NS, 100 mm, 13 mm
N° de catalogue WITG2.114.100
Fournisseur:   witeg Labortechnik
Drying tubes, U-shaped
N° de catalogue WITG2.114.100
Fournisseur:  witeg Labortechnik
Drying tubes, U-shaped
Drying tubes, U-shaped
12/21 NS, 100 mm, 13 mm
Prix
55,80 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Borosilicate glass, clear. Replaceable NS stopcocks. Lateral tube connectors.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Spécifications
  • Ø int.:
    13 mm
  • Rodage femelle:
    12/21 NS
  • Longueur:
    100 mm