Erlenmeyer flasks with screw caps
1000 ml, 220 mm, 131 mm, GL 32
N° de catalogue WITG5.507.006
Fournisseur: witeg Labortechnik
Aller à :
Product Details & Documents
Erlenmeyer flasks, made of borosilicate glass, with screw-thread, graduated can be used for the storage of culture media and mixing.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Spécifications
- Ø ext.:131 mm
- Filetage:GL 32
- Capacité:1000 ml
- Hauteur:220 mm