Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Molybdenum standard solution

Molybdenum standard solution

Molybdenum standard solution (NIST), 1000 mg/l as Mo, 100 mL
N° de catalogue HACH1418642
Fournisseur:   Hach
Molybdenum standard solution
N° de catalogue HACH1418642
Fournisseur:  Hach
Numéro CAS:  
Molybdenum standard solution
Molybdenum standard solution

Molybdenum standard solution (NIST), 1000 mg/l as Mo, 100 mL

Numéro CAS:  
Prix
38,00 €
Connexion Pour la disponibilité
1 * 100 mL
Product Details & Documents

Standard solution for accuracy checks in molybdenum determinations.

Spécifications

Spécifications

  • Concentration:
    1000 mg/l as Mo
  • Cdt:
    100 mL
  • Désignation:
    Molybdenum standard solution (NIST)

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.