Büchner funnels, Hartley pattern
Büchner funnels, Hartley pattern
90 mm
N° de catalogue 511-0838
Fournisseur:   DIXON SCIENTIFIC
Büchner funnels, Hartley pattern
N° de catalogue 511-0838
Fournisseur:  DIXON SCIENTIFIC
Büchner funnels, Hartley pattern
Büchner funnels, Hartley pattern
90 mm
Prix
435,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Top and base: Borosilicate glass with support plate.
  • Available with choice of perforated disc material
  • Complete with three PE coated clips
  • Available for 4 filter sizes
Three piece construction.
Spécifications
  • Pour filtre Ø:
    90 mm