Separating funnels, pear shaped
250 ml, 24/29 NS
N° de catalogue 511-0755
Fournisseur:   SciLabware
Separating funnels, pear shaped
N° de catalogue 511-0755
Fournisseur:  SciLabware
Separating funnels, pear shaped
Separating funnels, pear shaped
250 ml, 24/29 NS
Prix
94,60 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
PYREX®, borosilicate glass.
  • Without graduation
  • Very good chemical resistance
  • Very good temperature resistance
Available with ground glass stopper and interchangeable PTFE key.
Spécifications
  • Rodage femelle:
    24/29 NS
  • Capacité:
    250 ml