Powder funnels, Quickfit®
Powder funnels, Quickfit®
19/26 NS, 105 mm
N° de catalogue 201-8295
Fournisseur:   SciLabware
Powder funnels, Quickfit®
N° de catalogue 201-8295
Fournisseur:  SciLabware
Powder funnels, Quickfit®
Powder funnels, Quickfit®
19/26 NS, 105 mm
Prix
42,50 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
PYREX®, borosilicate glass, stem with cone.
  • Very good chemical resistance
  • High temperature resistance
Designed for transfer of powders into flasks and vessels.
Spécifications
  • Rodage mâle:
    19/26 NS
  • Ø ext. entonnoir sup.:
    105 mm
  • Seq:
    02