Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
 

Flasks, round bottom, Quickfit® ground joints

10 ml, 10/19 NS, 68 mm
  201-8466
 :   SciLabware
Flasks, round bottom, Quickfit® ground joints
  201-8466
Flasks, round bottom, Quickfit® ground joints
Flasks, round bottom, Quickfit® ground joints

10 ml, 10/19 NS, 68 mm

 
70,20 €
   
1 * 5 ST
 

PYREX®, borosilicate glass, clear.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

With white, matt labelling area.

 : ISO 1773, ISO 4797.

 
  • Rodage femelle:
    10/19 NS
  • Capacité:
    10 ml
  • Hauteur:
    68 mm
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.