BlocStor™ tissue storage boxes
BlocStor™ tissue storage boxes
Tissue storage boxes
N° de catalogue 720-2358
Fournisseur:   CELLPATH
BlocStor™ tissue storage boxes
N° de catalogue 720-2358
Fournisseur:  CELLPATH
BlocStor™ tissue storage boxes
BlocStor™ tissue storage boxes
Tissue storage boxes
Prix
146,00 €
Connexion Pour la disponibilité
1 * 25 ST
Product Details & Documents
Boxes from recycled cardboard.
  • Strong but light
  • With pre-printed archive label for identification
Livraison: Supplied in flat packs.
Spécifications
  • Désignation:
    Tissue storage boxes
  • Seq:
    02