Pipette tips, Ovation®
Standard, 35 µl, -, 10 portoirs, 96 chacun
N° de catalogue 613-4928
Fournisseur: VISTALAB TECHNOLOGIES
Aller à :
Product Details & Documents
Ideal for use with Ovation and MLA pipettes.
- Made of 100% pure virgin PP
- Provide a perfect fit
- Deliver reliable performance
- Autoclavable
Spécifications
- Emballage:10 portoirs, 96 chacun
- Stérile:-
- Volume:35 µl
- Seq:04
- Style de pointe:Standard