D-(+)-Cellobiose octaacetate
Glass bottle for solids, 5 g
Fournisseur: Thermo Fisher Scientific
D-(+)-Cellobiose octaacetate
N° de catalogue L08780.06
Fournisseur: Thermo Fisher Scientific
Numéro CAS: 5346-90-7
Aller à :
Product Details & Documents
CAS No.: 5346-90-7 Molecular Formula: C28H38O19 Formula Weight: 678.59 Melting Point: 224-231° Beilstein Reference: 79280 Merck Reference: 14,1961 Optical Rotation: +40° (c=1 in chloroform) MDL No.: MFCD00009600
- Numéro CAS:5346-90-7
- Formule:C₂₈H₃₈O₁₉
- Numéro MDL:MFCD00009600
- Poids moléculaire:678.59 g/mol
- Index Merck:14,1961
- Point de fusion:224…231 °C
- Température de stockage:Température ambiante
- EINECS:226-304-5
Spécifications
Spécifications
- Type de conditionnement:Glass bottle for solids
- Cdt:5 g
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



